missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.1142 Normal, Reagents

Sulfuric Acid, 0.1142 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

$162.83 - $590.70

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-333-887
View Documents
Labchem Inc
CS10030020F
20 L
Each for $590.70
Only null left
 
01-333-886
View Documents
Labchem Inc
CS1003004A
4 L
Each for $162.83
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Sulphuric Acid, Dihydrogen Sulfate
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

<2
7732-18-5,7664-93-9
Cubitainer
20 L
Liquid
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
Colorless
100°C
1.0036
H2O
Sulphuric Acid, Dihydrogen Sulfate
O
18.02
Sulfuric Acid, 0.1142 Normal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'