missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.0833 N (N/12), Ricca Chemical
For Preparation of Protein-free Filtrates
Supplier: Ricca Chemical Company 823032
Specifications
| Sulfuric Acid | |
| 98.0,0.42 | |
| Natural Poly Bottle | |
| H2O4S | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 |
| 7664-93-9,7732-18-5 | |
| 100.0,0.43 | |
| 1 L | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(=O)(=O)O | |
| 98.072 | |
| CHEBI:26836 |
Chemical Identifiers
| 7664-93-9 | |
| 98.072 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(=O)(=O)O |
| H2O4S | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |