Please login to your online account to display your discounted pricing

Sulfuric Acid, 0.0833 Normal, for Preparation of Protein-free Filtrates, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7664-93-9
CAS Min % 0.42
CAS Max % 0.43
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

8230-1

 
ricca chemical company
RI82301
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

8230-32

 
ricca chemical company
RI823032
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R823000020F

 
ricca chemical company
RIR823000020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Sulfuric Acid
CAS 7732-18-5
Concentration or Composition Notes 0.0833Normal
CAS Max % 100.0
Packaging Poly natural
CAS Max % 0.43
CAS Min % 0.42
CAS 7664-93-9
CAS Min % 98.0

Preparation of Protein-free Filtrates