missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.0500 N (N/20), Ricca Chemical

$54.63 - $272.88

Chemical Identifiers

CAS 7732-18-5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R8219000500
View Documents
Ricca Chemical Company
R8219000500A
500 mL Natural Poly Bottle
Each for $54.63
Only null left
 
8219-32
View Documents
Ricca Chemical Company
821932
1 L Natural Poly Bottle
Each for $110.44
Only null left
 
8219-1
View Documents
Ricca Chemical Company
82191
4 L Natural Poly Bottle
Each for $272.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5

Specifications

Colorless
98.0,0.25
100.0,0.26
500 mL
Liquid
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Sulfuric Acid
7732-18-5,7664-93-9
0.0500Normal
Natural Poly Bottle
H2O4S
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Laboratory
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'