missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.040 Normal, Reagents
Sulfuric Acid, 0.040 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
$72.27 - $296.40
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Sulphuric Acid, Dihydrogen Sulfate |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-333-713
|
Labchem Inc
CS1332001A |
1 L |
Each for $90.33
|
|
|||||
|
01-333-712
|
Labchem Inc
CS1332004A |
4 L |
Each for $296.40
|
|
|||||
|
01-333-711
|
Labchem Inc
CS133200500A |
500 mL |
Each for $72.27
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| <2 | |
| 7732-18-5,7664-93-9 | |
| Poly Bottle | |
| 1 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
| Colorless | |
| 100°C | |
| 1.0013 | |
| H2O | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O | |
| 18.02 | |
| Sulfuric Acid, 0.040 Normal |