missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.0200 N (N/50), Ricca Tightrant™

Catalog No. 82001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
82001 4 L Natural Poly Bottle
820016 500 mL Natural Poly Bottle
820032 1 L Natural Poly Bottle
820025 10 L Cubitainer
82005 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 82001 Supplier Ricca Chemical Company Supplier No. 82001
Edge
  • Tested in and ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Packaging Natural Poly Bottle
Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0200N
Chemical Name or Material Sulfuric Acid