missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.015 Normal, Reagents

Catalog No. 01333616
Change view
Click to view available options
Quantity: 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01333616 Quantity: 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333616 Supplier Labchem Inc Supplier No. CS14660010F
Only null left

Sulfuric Acid, 0.015 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH 2.12
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0005
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 0.015 Normal