missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.0100 Normal, Reagents

Catalog No. 01336372
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336372 1 L
01336371 20 L
01336370 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336372 Supplier Labchem Inc Supplier No. C73053501A

Sulfuric Acid, 0.0100 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH 2.3
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0003
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 0.0100 Normal