missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Specifications
| Melting Point | 155.0°C to 158.0°C |
| Color | Beige to Brown |
| Flash Point | >120°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Linear Formula | SO3·HCON(CH3)2 |
| Quantity | 5 g |
| Formula Weight | 153.16 |
| Percent Purity | 47+% active SO3 |
| Physical Form | Crystals |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with wat |
| EINECSNumber | 249-701-5 |
RUO – Research Use Only