accessibility menu, dialog, popup

UserName

CH3NH3COOSOO

CAS RN 29584-42-7

IUPAC Name: N,N-dimethylformamide; sulfonylideneoxidane
Synonyms: dimethylformamide; sulfur trioxide
Molecular Weight (g/mol): 153.15
Molecular Formula: C3H7NO4S
InChi Key: AFDQGRURHDVABZ-UHFFFAOYSA-N
SMILES: O=S(=O)=O.CN(C)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results