missing translation for 'onlineSavingsMsg'
Learn More

Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3

Catalog No. AC425850050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425850050 5 g Glass bottle
AC425850250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425850050 Supplier Thermo Scientific Chemicals Supplier No. 425850050
Only null left

CAS: 29584-42-7 | C3H7NO4S | 153.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Chemical Identifiers

CAS 29584-42-7
Molecular Formula C3H7NO4S
Molecular Weight (g/mol) 153.15
MDL Number MFCD00043411
InChI Key AFDQGRURHDVABZ-UHFFFAOYSA-N
Synonym n,n-dimethylformamide sulfur trioxide complex, dmf-sulfur trioxide complex, n,n-dimethylformamide; sulfur trioxide, sulfur trioxide dimethylformamide complex, formamide, n,n-dimethyl-, compd. with sulfur trioxide 1:1, dmf.so3, n,n-dimethylformamide, compound with sulphur trioxide, dimethyl formamide sulfur trioxide, dimethylformamide; sulfur trioxide, sulfur trioxide n,n-dimethylformamide complex
PubChem CID 169056
IUPAC Name N,N-dimethylformamide;sulfur trioxide
SMILES O=S(=O)=O.CN(C)C=O

Specifications

Melting Point 155.0°C to 158.0°C
Color Beige to Brown
Flash Point >120°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula SO3·HCON(CH3)2
Quantity 5 g
Formula Weight 153.16
Percent Purity 47+% active SO3
Physical Form Crystals
Chemical Name or Material Sulfur trioxide N, N-dimethylformamide complex, 47+% active SO3
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 249-701-5

RUO – Research Use Only