missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Kerosene standard solution, Specpure™, 100μg/g (0.010%)

Catalog No. AA40537AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40537AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40537AE Supplier Thermo Scientific Chemicals Supplier No. 040537AE
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00804268

Specifications

UN Number UN1223
Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Kerosene standard solution, 100μg/g (0.010%)
Hazard Category H227-H304
Hazard Statement GHS H Statement
H304-H227
May be fatal if swallowed and enters airways.
Combustible liquid.
Precautionary Statement P210-P280-P301+P310-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KEROSENE
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only