missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur ICP Standard, 10,000 ppm S in H2O, Ricca Chemical
Chemical Identifiers
| CAS | 7783-20-2 , 7732-18-5 |
|---|---|
| Molecular Formula | H8N2O4S |
| Molecular Weight (g/mol) | 132.134 |
| InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
| PubChem CID | 6097028 |
| ChEBI | CHEBI:62946 |
| IUPAC Name | diazanium;sulfate |
| SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)[O-] |
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | dimQuantity | Packaging | Price | Quantity & Availability | ||||
|
PS10KW50
|
N/A | Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
PS10KW100
|
N/A | Natural LDPE Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
PS10KW500
|
N/A | Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| H8N2O4S | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| CHEBI:62946 | |
| [NH4+].[NH4+].[O-]S(=O)(=O)[O-] |
Specifications
| 7783-20-2 , 7732-18-5 | |
| 94.0, 3.88 | |
| 1 mL = 10 mg S | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| diazanium;sulfate | |
| 6097028 | |
| Sulfur ICP Standard |
| 10,000ppm S | |
| 98.0, 4.04 | |
| H8N2O4S | |
| [NH4+].[NH4+].[O-]S(=O)(=O)[O-] | |
| 132.134 | |
| CHEBI:62946 |