missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur in Heavy Mineral Oil standard solution, Specpure™, 3000μg/g (0.30%)
Supplier: Thermo Scientific Chemicals 040353AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sulfur in Heavy Mineral Oil standard solution | |
| MFCD00804270 | |
| Specpure |
| 3000μg/g (0.30%) | |
| 100 mL |
Chemical Identifiers
| MFCD00804270 |
Safety and Handling
GHS H Statement
H350
May cause cancer.
P301+P310-P331-P501c
H304-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only