missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Heavy Mineral Oil standard solution, Specpure™, 10,000μg/g (1.00%)

Catalog No. AA40356AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40356AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40356AE Supplier Thermo Scientific Chemicals Supplier No. 040356AE
Only null left

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00804270

Specifications

Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Heavy Mineral Oil standard solution, 10,000μg/g (1.00%)
Hazard Category H304-H335
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P301+P310-P331-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only