missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur in Diesel Fuel standard solution, Specpure™, 500μg/g (0.050%)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 100 mL |
| Grade | Specpure |
| Chemical Name or Material | Sulfur in Diesel Fuel standard solution, 500μg/g (0.050%) |
Safety and Handling
| Hazard Category | H227-H351 |
| Hazard Statement | GHS H Statement H351-H315-H319-H335-H227 Suspected of causing cancer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P201-P202-P210-P281-P308+P313-P370+P378q-P501c |
| EINECSNumber | 270-676-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only