missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Diesel Fuel standard solution, Specpure™, 10,000μg/g (1.00%)

 

Supplier:  Thermo Scientific Chemicals 038776AE

Catalog No. AA38776AE


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sulfur in Diesel Fuel standard solution
MFCD01095233
Specpure
10,000μg/g (1.00%)
100 mL

Chemical Identifiers

MFCD01095233
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P201-P202-P210-P235-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335-H351

EINECSNumber : 270-676-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only