missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Crude Oil standard solution, Specpure™, 20,000μg/g (2.00%)

Catalog No. AA41889AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41889AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41889AE Supplier Thermo Scientific Chemicals Supplier No. 041889AE

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00197838

Specifications

Odor Unpleasant
Physical Form Liquid
UN Number UN1267
Color Black
Quantity 100 mL
Grade Specpure
Chemical Name or Material Sulfur in Crude Oil standard solution, 20,000μg/g (2.00%)
Hazard Category H226-H335-H350
Hazard Statement GHS H Statement
H350-H226
May cause cancer.
Flammable liquid and vapor.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PETROLEUM CRUDE OIL
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only