missing translation for 'onlineSavingsMsg'
Learn More

Sulfur in Crude Oil standard solution, 1000μg/g (0.10%), Specpure™

Catalog No. AA41885AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41885AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41885AE Supplier Thermo Scientific Chemicals Supplier No. 041885AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7704-34-9 | S | 32.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7704-34-9
Molecular Formula S
Molecular Weight (g/mol) 32.06
MDL Number MFCD00085316
InChI Key NINIDFKCEFEMDL-UHFFFAOYSA-N
PubChem CID 5362487
ChEBI CHEBI:26833
IUPAC Name sulfur
SMILES [S]

Specifications

Odor Unpleasant
Physical Form Liquid
Color Black
Quantity 100 mL
Formula Weight 3350
Chemical Name or Material Sulfur, 1000μg/g (0.10%)
Hazard Category H226-H350
Hazard Statement Avoid exposure - obtain special instructions before use. When using do not eat or drink. Wear suitable protective clothing and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only