missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur AA Standard, 1000 ppm S in H2O, Ricca Chemical
$132.84 - $329.47
Chemical Identifiers
| CAS | 7783-20-2 |
|---|---|
| Molecular Formula | H8N2O4S |
| Molecular Weight (g/mol) | 132.13 |
| MDL Number | MFCD00003391 |
| InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
| PubChem CID | 6097028 |
| ChEBI | CHEBI:62946 |
| IUPAC Name | sulfuric acid diamine |
| SMILES | N.N.OS(O)(=O)=O |
Chemical Identifiers
| 7783-20-2 | |
| 132.13 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| CHEBI:62946 | |
| N.N.OS(O)(=O)=O |
| H8N2O4S | |
| MFCD00003391 | |
| 6097028 | |
| sulfuric acid diamine |
Specifications
| Natural LDPE Bottle | |
| 98.0,0.4 | |
| Liquid | |
| MFCD00003391 | |
| 100°C | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| sulfuric acid diamine | |
| 100 mL | |
| CHEBI:62946 | |
| Sulfur AA Standard |
| 7783-20-2,7732-18-5 | |
| 100.0,0.42 | |
| H8N2O4S | |
| Colorless | |
| Standard | |
| N.N.OS(O)(=O)=O | |
| 132.13 | |
| 6097028 | |
| 1mL = 1mg S (1, 000ppm S)(NH4)2SO4 in H2O |
Safety and Handling
ShelfLife : 24 months