missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, 3% (w/v), Ricca Chemical

Supplier:  Ricca Chemical Company 81151

 View more versions of this product

Catalog No. 8115-1

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfosalicylic Acid Solution
C7H6O6S
YCPXWRQRBFJBPZ-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid
7322
3%
3% (w/v)
Aqueous solution
97-05-2
5-sulfosalicylic acid, sulfosalicylic acid, 5-sulphosalicylic acid, salicylsulfonic acid, benzoic acid, 2-hydroxy-5-sulfo, salicylic acid, sulfo, benzoic acid, 2-hydroxysulfo, sulphosalicylic acid, salicylic acid, 5-sulfo, 2-hydroxybenzoic-5-sulfonic acid
C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O
218.179
CHEBI:68555
Laboratory
Amber Glass Bottle
4 L

Chemical Identifiers

97-05-2
218.179
5-sulfosalicylic acid, sulfosalicylic acid, 5-sulphosalicylic acid, salicylsulfonic acid, benzoic acid, 2-hydroxy-5-sulfo, salicylic acid, sulfo, benzoic acid, 2-hydroxysulfo, sulphosalicylic acid, salicylic acid, 5-sulfo, 2-hydroxybenzoic-5-sulfonic acid
CHEBI:68555
C1=CC(=C(C=C1S(=O)(=O)O)C(=O)O)O
C7H6O6S
YCPXWRQRBFJBPZ-UHFFFAOYSA-N
7322
2-hydroxy-5-sulfobenzoic acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions