missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulforhodamine 101 (free acid), 99%, pure, laser grade

Catalog No. AC419941000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419941000 100 mg
AC419945000 500 mg
AC419940025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419941000 Supplier Thermo Scientific Chemicals Supplier No. 419941000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60311-02-6 | C31H29ClN2O6S2 | 625.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 60311-02-6
Molecular Formula C31H29ClN2O6S2
Molecular Weight (g/mol) 625.15
MDL Number MFCD00012407
InChI Key MPLHNVLQVRSVEE-UHFFFAOYSA-N
Synonym sulforhodamine 101, sulforhodamine 101; sulforhodamine 640, unii-fx0es3271v, texas red, sulforhodamine 101 acid chloride, 2-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-pyrido 3,2,1-ij quinolizino 1',9':6,7,8 chromeno 2,3-f quinolin-4-ium-9-yl-5-sulfobenzenesulfonate, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9$l^ 5 ,23-diazaheptacyclo 17.7.1.1^ 5,9 .0^ 2,17 .0^ 4,15 .0^ 23,27 .0^ 13,28 octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9??,23-diazaheptacyclo 17.7.1.1?,?.0 2 , 1 ?.0?, 1 ?.0 2 3 , 2 ?.0 1 3 , 2 ? octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9lambda,23-diazaheptacyclo 17.7.1.1,.0 2 , 1 .0, 1 .0 2 3 , 2 .0 1 3 , 2 octacosa-1,4,9 28 ,13,15,17,19 27-heptaen-9-ylium, 9-2,4-disulfophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium inner salt, hydrogen 9-2,4-disulphonatophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium
PubChem CID 122180
ChEBI CHEBI:52274
IUPAC Name 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium
SMILES [O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O

Specifications

Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 606.71
Percent Purity 99%
Grade Pure
Chemical Name or Material Sulforhodamine 101 (free acid), (free acid)
EINECSNumber 262-159-4

RUO – Research Use Only