missing translation for 'onlineSavingsMsg'
Learn More

Chiron Sulfonylglycine Standards

Catalog No. 14957850me
Change view
Click to view available options
Chemical Name or Material:
N-Perfluorobutanesulfonylglycine
N-Perfluorohexanesulfonylglycine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
14957850ME N-Perfluorohexanesulfonylglycine
14901650ME N-Perfluorobutanesulfonylglycine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14957850ME Supplier Chiron Supplier No. 14957.850ME
Only null left

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material N-Perfluorohexanesulfonylglycine
CAS 1003193-99-4
Concentration 50 μg/mL
Synonym FHxSAA
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL