missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chiron Sulfonylglycine Standards
Description
- Various singles available
- Solutions in assorted solvents
- Useful for various matrices
- Custom standards of other PFAS compounds available on demand
Specifications
Specifications
| Chemical Name or Material | N-Perfluorobutanesulfonylglycine |
| CAS | 347872-22-4 |
| Concentration | 50 μg/mL |
| Synonym | FBSAA |
| Type | Reference Material |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |