missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chiron Sulfonylglycine, N-methyl Standards
PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.
$2,146.61 - $2,299.61
Specifications
| Concentration | 50 μg/mL |
|---|---|
| Type | Reference Material |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Catalog Number | Mfr. No. | Chemical Name or Material | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Chemical Name or Material | Price | Quantity | |||||
|
14-949-750ME
|
Chiron
14949750ME |
N-Methyl-n-perfluorobutanesulfonylglycine-d3 |
N/A
|
|
|||||
|
14-950-950ME
|
Chiron
14950950ME |
N-Methyl-n-perfluorohexanesulfonylglycine |
N/A
|
|
|||||
|
14-951-950ME
|
Chiron
14951950ME |
N-Methyl-n-perfluorohexanesulfonylglycine-d3 |
N/A
|
|
|||||
Description
- Various singles available
- Solutions in assorted solvents
- Useful for various matrices
- Custom standards of other PFAS compounds available on demand
Specifications
| 50 μg/mL | |
| Reference Material | |
| Methanol | |
| 1 mL |