missing translation for 'onlineSavingsMsg'
Learn More

Chiron Sulfonic Acid Standards

Catalog No. 15140250ME
Change view
Click to view available options
Chemical Name or Material:
Perfluorododecanesulfonic acid
Perfluoroethanesulfonic acid, potassium salt
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
15140250ME Perfluoroethanesulfonic acid, potassium salt
149631250ME Perfluorododecanesulfonic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15140250ME Supplier Chiron Supplier No. 15140.250ME
Only null left

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material Perfluoroethanesulfonic acid, potassium salt
CAS 2837-92-5
Concentration 50 μg conj. base/mL
Synonym PFES-K
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL