missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfathiazole 98.0+%, TCI America™
Supplier: TCI America S027225G
Specifications
| Sulfathiazole | |
| 200°C | |
| C9H9N3O2S2 | |
| 25 g | |
| JNMRHUJNCSQMMB-UHFFFAOYSA-N | |
| 4-amino-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide | |
| 5340 | |
| 255.31 | |
| Crystalline Powder |
| 72-14-0 | |
| White-Yellow | |
| MFCD00005319 | |
| sulfathiazole, sulphathiazole, sulfathiazol, sulfanilamidothiazole, 2-sulfanilamidothiazole, norsulfazole, thiazamide, 2-sulfonamidothiazole, norsulfasol, neostrepsan | |
| NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1 | |
| 255.31 | |
| CHEBI:9337 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 72-14-0 | |
| 255.31 | |
| JNMRHUJNCSQMMB-UHFFFAOYSA-N | |
| 5340 | |
| 4-amino-N-(1,3-thiazol-2-yl)benzene-1-sulfonamide |
| C9H9N3O2S2 | |
| MFCD00005319 | |
| sulfathiazole, sulphathiazole, sulfathiazol, sulfanilamidothiazole, 2-sulfanilamidothiazole, norsulfazole, thiazamide, 2-sulfonamidothiazole, norsulfasol, neostrepsan | |
| CHEBI:9337 | |
| NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1 |
Safety and Handling
RTECSNumber : WP2360000
TSCA : Yes