missing translation for 'onlineSavingsMsg'
Learn More

Sulfathiazole, 98+%

Catalog No. p-7080962
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1072722 100 g
AAA1072736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1072722 Supplier Thermo Scientific Chemicals Supplier No. A1072722

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72-14-0 | C9H9N3O2S2 | 255.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Mixture of hexanes where n-hexane content is ≥60%
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) and tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only

Chemical Identifiers

CAS 72-14-0
Molecular Formula C9H9N3O2S2
Molecular Weight (g/mol) 255.31
MDL Number MFCD00005319
InChI Key JNMRHUJNCSQMMB-UHFFFAOYSA-N
Synonym sulfathiazole, sulphathiazole, sulfathiazol, sulfanilamidothiazole, 2-sulfanilamidothiazole, norsulfazole, thiazamide, 2-sulfonamidothiazole, norsulfasol, neostrepsan
PubChem CID 5340
ChEBI CHEBI:9337
IUPAC Name 4-amino-N-(1,3-thiazol-2-yl)benzenesulfonamide
SMILES NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1

Specifications

Melting Point 200°C to 203°C
Assay Percent Range 99%
Quantity 100 g
Merck Index 14,8943
Solubility Information Slightly soluble in water,deuterated dimethyl sulfoxide and ethanol. Soluble in acetone,dilute mineral acids,potassium hydroxide solution,sodium hydroxide solution,ammonia water,dimethyl sulphoxide and methanol.
Formula Weight 255.32
Percent Purity 99%
Chemical Name or Material Sulfathiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-771-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only