missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate Standard, 100 ppm SO42-, Ricca Chemical
Supplier: Ricca Chemical Company 811016
Specifications
| Sulfate Standard | |
| 7757-82-6,7732-18-5 | |
| 100.0,0.02 | |
| Liquid | |
| MFCD00003504 | |
| 100°C | |
| [Na+].[Na+].[O-]S([O-])(=O)=O | |
| 142.04 | |
| 24436 |
| Natural Poly Bottle | |
| 98.0,0.01 | |
| 1 mL = 0.1 mg SO | |
| Na2O4S | |
| Colorless | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| disodium sulfate | |
| 500 mL | |
| CHEBI:32149 |
Chemical Identifiers
| 7757-82-6 | |
| 142.04 | |
| PMZURENOXWZQFD-UHFFFAOYSA-L | |
| CHEBI:32149 | |
| [Na+].[Na+].[O-]S([O-])(=O)=O |
| Na2O4S | |
| MFCD00003504 | |
| 24436 | |
| disodium sulfate |