missing translation for 'onlineSavingsMsg'
Learn More

Sulfate, Ion chromatography standard solution, Specpure™, SO4-2 1000μg/mL

$248.96 - $452.64

Chemical Identifiers

MDL Number MFCD00145343
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35547AE
View Documents
Thermo Scientific Chemicals
035547AE
100 mL
Each for $248.96
Only null left
 
AA35547AP
View Documents
Thermo Scientific Chemicals
035547AP
500 mL
Each for $452.64
Only null left
 
Description

Description

Sulfate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is also used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00145343

Specifications

Liquid
Miscible with water
Specpure
MFCD00145343
100 mL
Sulfate Ion chromatography standard solution, SO4-2 1000μg/mL
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only