missing translation for 'onlineSavingsMsg'
Learn More

Sulfate, Ion chromatography standard solution, Specpure™, SO4-2 1000μg/mL

Catalog No. AA35547AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35547AP 500 mL
AA35547AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35547AP Supplier Thermo Scientific Chemicals Supplier No. 035547AP
Only null left

Sulfate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is also used for calibration of ion chromatography and other analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00145343

Specifications

Physical Form Liquid
Solubility Miscible with water
Quantity 500 mL
Grade Specpure
Chemical Name or Material Sulfate Ion chromatography standard solution, SO4-2 1000μg/mL
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only