missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfate, Ion chromatography standard solution, Specpure™, SO4-2 1000μg/mL
Description
Sulfate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is also used for calibration of ion chromatography and other analytical techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Solubility | Miscible with water |
| Quantity | 100 mL |
| Grade | Specpure |
| Chemical Name or Material | Sulfate Ion chromatography standard solution, SO4-2 1000μg/mL |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only