missing translation for 'onlineSavingsMsg'
Learn More

Sulfapyridine melting point standard Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013128
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013128 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013128 Supplier Merck Emd Millipore Supplier No. PHR10871G

Sulfapyridine Melting Point Standard also called Sulfapyridine is a metabolite of Sulfasalazine. It is an antibiotic and is mostly used in treating rheumatoid arthritis.

Pharmaceutical (small molecule)

Specifications

CAS 144-83-2
Physical Form Neat
Linear Formula C11H11N3O2 S
MDL Number MFCD00038036
Melting Point ∼191°C
Synonym Sulfapyridine; 4-Amino-N-[2-pyridyl]benzene sulfonamide; N1-(Pyridin-2-yl)sulfanilamide
Type Certified Reference Material
Quantity 1 g
Formula Weight 249.29
Grade Certified Reference Material