missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfapyridine, ≥99.0%, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 144-83-2 |
| Packaging | Poly Bottle |
| Linear Formula | C11H11N3O2 S |
| Quantity | 25 g |
| Formula Weight | 249.29 |
| Percent Purity | ≥99.0% |
| Physical Form | Neat |