missing translation for 'onlineSavingsMsg'
Learn More

Sulfapyridine, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111013129
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013129 25 g
111013130 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013129 Supplier Merck Emd Millipore Supplier No. S625225G

Sulfapyridine is a metabolite of Sulfasalazine. It is an antibiotic and is mostly used in treating rheumatoid arthritis.

Pharmaceutical (small molecule)

Chemical Identifiers

MDL Number MFCD00038036
Synonym 4-Amino-N-[2-pyridyl]benzene sulfonamide; N1-(Pyridin-2-yl)sulfanilamide

Specifications

CAS 144-83-2
Packaging Poly Bottle
Linear Formula C11H11N3O2 S
Quantity 25 g
Formula Weight 249.29
Percent Purity ≥99.0%
Physical Form Neat