missing translation for 'onlineSavingsMsg'
Learn More

Sulfamide 96.0+%, TCI America™

Catalog No. p-7135981
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
250 g
Catalog No. Quantity
S089025G 25 g
S0890250G 250 g
2 options

Catalog No. S089025G

Supplier: TCI America S089025G

Only null left

Chemical Identifiers

CAS 7803-58-9
Molecular Formula H4N2O2S
Molecular Weight (g/mol) 96.10
MDL Number MFCD00011606
InChI Key NVBFHJWHLNUMCV-UHFFFAOYSA-N
Synonym sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid
PubChem CID 82267
ChEBI CHEBI:29368
IUPAC Name sulfamoylamine
SMILES NS(N)(=O)=O

Specifications

Melting Point 91°C
Color White
Quantity 25 g
Formula Weight 96.10
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Sulfamide