missing translation for 'onlineSavingsMsg'
Learn More

Sulfamide, 99%

Catalog No. p-7024291
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1868088 2.5 g
AAA1868009 10 g
AAA1868018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1868088 Supplier Thermo Scientific Chemicals Supplier No. A1868088
Only null left

CAS: 7803-58-9 | H4N2O2S | 96.10 g/mol

It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

Solubility
Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol

Notes
Moisture Sensitive. Store away from oxidizing agents, water/humidity. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 7803-58-9
Molecular Formula H4N2O2S
Molecular Weight (g/mol) 96.10
MDL Number MFCD00011606
InChI Key NVBFHJWHLNUMCV-UHFFFAOYSA-N
Synonym sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid
PubChem CID 82267
ChEBI CHEBI:29368
SMILES NS(N)(=O)=O

Specifications

Melting Point 91°C to 92°C
Density 1.61
Quantity 2.5 g
Sensitivity Moisture sensitive
Merck Index 14,8922
Solubility Information Soluble in water, hot alcohol, acetone; Sparingly soluble in cold alcohol
IUPAC Name sulfamide
Formula Weight 96.11
Percent Purity 99%
Chemical Name or Material Sulfamide
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-262-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only