missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3339998
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
White Poly Wide Mouth Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RDCS1100100 100 g White Poly Wide Mouth Bottle
RDCS1100500 500 g White Poly Wide Mouth Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RDCS1100100 Supplier Ricca Chemical Company Supplier No. RDCS1100100B1
Only null left

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Heavy Metals (as Pb) max. 0.001%
Packaging White Poly Wide Mouth Bottle
Residue after Ignition max. 0.01%
Quantity 100 g
Physical Form Crystals
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
SMILES NS(=O)(=O)O
Molecular Weight (g/mol) 97.088
Percent Purity 99.3 to 100.3%
Grade ACS
Chemical Name or Material Sulfamic Acid
Show More Show Less
ShelfLife 60 months