missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, ACS Reagent, Reagents

Catalog No. p-200020968
Change view
Click to view available options
Quantity:
100 mL
500 g
2.5 kg
6 lb.
10 kg
50 lb.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337575 100 mL
01337574 10 kg
01337573 2.5 kg
01337572 500 g
01337571 50 lb.
01337570 6 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01337575 Supplier Labchem Inc Supplier No. C2114300100A6
Only null left

Sulfamic Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.09
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid
IUPAC Name sulfamic acid
SMILES NS(O)(=O)=O

Specifications

Color White to Off-White
Melting Point 205°C
Packaging Poly Bottle
Specific Gravity 2.15
Quantity 100 mL
Physical Form Solid
Grade ACS
Chemical Name or Material Sulfamic Acid, ACS Reagent