missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, ACS Grade, 99.3 to 100.3%, LabChem™

Supplier:  LabChem LC254201

 View more versions of this product

Catalog No. LC254201

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfamic Acid
205°C
2,120kg/m3
Passes Test
2120kg/m3
500 g
Crystalline Powder
UN2967
Soluble in water
NS(=O)(=O)O
97.088
CHEBI:9330
99.3 to 100.3%
White
5329-14-6
100
Poly Bottle
Sulfur compounds
H3NO3S
NH2SO3H
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
IIACRCGMVDHOTQ-UHFFFAOYSA-N
sulfamic acid
5987
97.09
ACS

Chemical Identifiers

5329-14-6
97.088
amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
CHEBI:9330
NS(=O)(=O)O
H3NO3S
IIACRCGMVDHOTQ-UHFFFAOYSA-N
5987
sulfamic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Toxic to aquatic life.

GHS P Statement
Do not breathe vapors, mist, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or air): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 226-218-8

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions