missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic acid, ≥99.5% (alkalimetric), Honeywell™

Catalog No. p-7071737
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
6 x 2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001566 1 kg
6001567 2.5 kg
6001568 5 kg
60044601 6 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001566 Supplier Honeywell Chemicals Supplier No. 074011KG
Only null left

≥99.5% (alkalimetric)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
Molecular Weight (g/mol) 97.088
MDL Number MFCD00011603
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid
SMILES NS(=O)(=O)O

Specifications

Residue after Ignition ≤0.02%
Quantity 1 kg
Linear Formula NH2SO3H
UN Number 2967
Formula Weight 97.09g/mol
Percent Purity ≥99.5% (alkalimetric)
Chemical Name or Material Sulfamic Acid
Hazard Statement P273-P305 + P351 + P338
Precautionary Statement H315-H319-H412
EINECSNumber 226-218-8