missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic Acid, 10% w/v, Reagents
Sulfamic Acid, 10% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CS2100004A
Specifications
| Sulfamic Acid, 10% w/v | |
| 7732-18-5,5329-14-6 | |
| Poly Bottle | |
| 4 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 10% w/v |
| Colorless | |
| 100°C | |
| 1.05 | |
| H2O | |
| Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid | |
| O | |
| 18.02 | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Amidosulfonic Acid, Aminosulfonic Acid, Sulphamic Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |