missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxypyridazine 98.0+%, TCI America™
Supplier: TCI America S059125G
Specifications
| Sulfamethoxypyridazine | |
| 182°C | |
| C11H12N4O3S | |
| 25 g | |
| VLYWMPOKSSWJAL-UHFFFAOYSA-N | |
| 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide | |
| 5330 | |
| 280.30 | |
| Crystalline Powder |
| 80-35-3 | |
| Yellow | |
| MFCD00057372 | |
| sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine | |
| COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N | |
| 280.302 | |
| CHEBI:102516 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 80-35-3 | |
| 280.302 | |
| VLYWMPOKSSWJAL-UHFFFAOYSA-N | |
| 5330 | |
| 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide |
| C11H12N4O3S | |
| MFCD00057372 | |
| sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine | |
| CHEBI:102516 | |
| COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N |
Safety and Handling
RTECSNumber : WP0400000
TSCA : No