missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxypyridazine, 99.04%, MP Biomedicals™
$78.03 - $223.90
Chemical Identifiers
| CAS | 80-35-3 |
|---|---|
| Molecular Formula | C11H12N4O3S |
| Molecular Weight (g/mol) | 280.302 |
| InChI Key | VLYWMPOKSSWJAL-UHFFFAOYSA-N |
| Synonym | sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine |
| PubChem CID | 5330 |
| ChEBI | CHEBI:102516 |
| IUPAC Name | 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide |
| SMILES | COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15671201
|
MP Biomedicals Inc
0215671201 |
1 g |
Each for $78.03
|
|
|||||
|
ICN15671225
|
MP Biomedicals Inc
0215671225 |
25 g |
Each for $223.90
|
|
|||||
Chemical Identifiers
| 80-35-3 | |
| 280.302 | |
| sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine | |
| CHEBI:102516 | |
| COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N |
| C11H12N4O3S | |
| VLYWMPOKSSWJAL-UHFFFAOYSA-N | |
| 5330 | |
| 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide |
Specifications
| 80-35-3 | |
| 1 g | |
| VLYWMPOKSSWJAL-UHFFFAOYSA-N | |
| 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide | |
| 5330 | |
| 280.3 | |
| Crystalline |
| C11H12N4O3S | |
| sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine | |
| COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N | |
| 280.302 | |
| CHEBI:102516 | |
| 0.9904 | |
| Sulfamethoxypyridazine |
Safety and Handling
EINECSNumber : 201-272-5
Recommended Storage : Store at 2° to 8°C.