missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxypyridazine, 99.04%, MP Biomedicals™

Catalog No. ICN15671225
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15671225 25 g
ICN15671201 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15671225 Supplier MP Biomedicals Inc Supplier No. 0215671225

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80-35-3
Molecular Formula C11H12N4O3S
Molecular Weight (g/mol) 280.302
InChI Key VLYWMPOKSSWJAL-UHFFFAOYSA-N
Synonym sulfamethoxypyridazine, sulphamethoxypyridazine, sulfapyridazine, sulfalex, longin, midicel, sulfametoxipiridazine, spofadazine, sulfdurazin, 3-sulfa-6-methoxypyridazine
PubChem CID 5330
ChEBI CHEBI:102516
IUPAC Name 4-amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide
SMILES COC1=NN=C(C=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Quantity 25 g
Formula Weight 280.3
Percent Purity 0.9904
Physical Form Crystalline
Chemical Name or Material Sulfamethoxypyridazine
EINECSNumber 201-272-5
Recommended Storage Store at 2° to 8°C.