missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxypyridazine, 99.04%, MP Biomedicals™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 280.3 |
| Percent Purity | 0.9904 |
| Physical Form | Crystalline |
| Chemical Name or Material | Sulfamethoxypyridazine |
Safety and Handling
| EINECSNumber | 201-272-5 |
| Recommended Storage | Store at 2° to 8°C. |