missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxazole, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111015358
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015358 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015358 Supplier Merck Emd Millipore Supplier No. PHR11261G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards.Sulfamethoxazole is one of the most effective sulfa drugs used for treating urinary infections in humans.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Specifications

CAS 723-46-6
Physical Form Neat
Linear Formula C10H11N3O3S
MDL Number MFCD00010546
Synonym 4-Amino-N-(5-methyl-3-isoxazolyl)benzenesulfonamide; N1-(5-Methylisoxazol-3-yl)sulfanilamide
Quantity 1 g
Formula Weight 253.28
Grade Certified Reference Material