missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxazole, MilliporeSigma™ Supelco™
Sulfamethoxazole is a bacteriostatic antibiotic, belonging to the class of sulfonamides. It is widely used in the treatment of pneumocystis, coccidiosis, gastroenteritis, diarrhea, respiratory diseases such as pneumonia, etc.
$92.70 - $452.18
Specifications
| CAS | 723-46-6 |
|---|---|
| Physical Form | Neat |
| Linear Formula | C10H11N3O3 S |
| MDL Number | MFCD00010546 |
| Synonym | 4-Amino-N-(5-methyl-3-isoxazolyl)benzenesulfonamide; N1-(5-Methylisoxazol-3-yl)sulfanilamide |
Description
Pharmaceutical (small molecule)Specifications
| 723-46-6 | |
| C10H11N3O3 S | |
| 4-Amino-N-(5-methyl-3-isoxazolyl)benzenesulfonamide; N1-(5-Methylisoxazol-3-yl)sulfanilamide | |
| 253.28 |
| Neat | |
| MFCD00010546 | |
| Analytical Standard | |
| Analytical Standard |