missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulfamethazine sodium salt

Catalog No. AC465600050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465600050 5 g
AC465600250 25 g
AC465601000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC465600050 Supplier Thermo Scientific Chemicals Supplier No. 465600050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1981-58-4
Molecular Weight (g/mol) 300.31
MDL Number MFCD12352107
Synonym Sulfadimidine sodium salt

Specifications

Type Sulfamethazine Sodium Salt
Physical Form Crystalline Powder
Source Synthetic
Infrared Spectrum Conforms
Loss on Drying ≤2 %
Packaging Glass Bottle
Color White
Quantity 5 g
Molecular Formula C12 H13 N4 Na O2 S
Shipping Condition normal conditions
Solubility Information soluble aq. soln.
Formula Weight 300.31
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Sulfamethazine sodium salt
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 5 years
storageNote1 Refrigerator +4°C

RUO – Research Use Only