missing translation for 'onlineSavingsMsg'
Learn More

Sulfamerazine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012905
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012905 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012905 Supplier Merck Emd Millipore Supplier No. PHR15771G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Sulfamerazine belongs to the class of sulfonamides, used as an antibacterial agent to treat infections, especially for patients intolerant to antibiotics.

Specifications

CAS 127-79-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C11H12N4O2 S
MDL Number MFCD00023212
Synonym Sulfamerazine; 4-Amino-N-(4-methyl-2-pyrimidinyl)benzenesulfonamide; N1-(4-Methylpyrimidin-2-yl)sulfanilamide
Type Certified Reference Material
Quantity 1 g
Formula Weight 264.30
Grade Certified Reference Material